Documents

N114

Supelco

Nifedipine

Synonym(s):
1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester
Empirical Formula (Hill Notation):
C17H18N2O6
CAS Number:
Molecular Weight:
346.33
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

form

neat

application(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

COC(=O)C1=C(C)NC(C)=C(C1c2ccccc2[N+]([O-])=O)C(=O)OC

InChI

1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18H,1-4H3

InChI key

HYIMSNHJOBLJNT-UHFFFAOYSA-N

Gene Information

human ... CACNA1C(775), CACNA1D(776), CACNA1F(778), CACNA1S(779)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

WGK Germany

1

Certificate of Analysis
Certificate of Origin
Joel S Klinck et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 157(2), 236-250 (2012-12-04)
Transport of calcium (Ca) and cadmium (Cd) was examined along the gastro-intestinal tract (GIT) of freshwater and seawater Oncorhynchus mykiss irideus (FWT and SWTies respectively) using in vitro and in vivo experiments. Based on known physiological differences between FWT and...
Carolien Roos et al.
JAMA, 309(1), 41-47 (2013-01-03)
In threatened preterm labor, maintenance tocolysis with nifedipine, after an initial course of tocolysis and corticosteroids for 48 hours, may improve perinatal outcome. To determine whether maintenance tocolysis with nifedipine will reduce adverse perinatal outcomes due to premature birth. APOSTEL-II...
Luc Desfrere et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(7), 2212-2217 (2009-01-24)
Dendritic growth is pivotal in the neurogenesis of cortical neurons. The sodium pump, or Na,K-ATPase, is an evolutionarily conserved protein that, in addition to its central role in establishing the electrochemical gradient, has recently been reported to function as a...
Yui Yamamoto et al.
Journal of pharmacological sciences, 121(3), 212-226 (2013-03-02)
We recently developed a novel cognitive enhancer, ST101 (spiro[imidazo[1,2-a]pyridine-3,2-indan]-2(3H)-one), that activates T-type voltage-gated calcium channels (VGCCs). Here, we address whether T-type VGCC activation with ST101 mediates its cognitive effects in vivo and the relevance of T-type VGCC activation to acetylcholine...
C B Wilcox et al.
BJOG : an international journal of obstetrics and gynaecology, 118(4), 423-428 (2011-01-05)
The success rates of external cephalic version (ECV) are improved with the use of betamimetic tocolytics, but these drugs are associated with maternal side effects. To critically evaluate the effectiveness and advantages, if any, of nifedipine as a tocolytic for...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.