Documents

N1765

Sigma-Aldrich

Monoclonal Anti-human Neutrophils, clone pmn 13F6

whole antiserum

MDL number:
Pricing and availability is not currently available.

antibody form

whole antiserum

antibody product type

primary antibodies

conjugate

unconjugated

shipped in

dry ice

storage temp.

−20°C

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.