Skip to Content
MilliporeSigma

N3273

2-Nitrophenyl-N-acetyl-β-D-galactosaminide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
UNSPSC Code:
12352204
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-5-3-2-4-8(9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)

SMILES string

CC(=O)NC1C(O)C(O)C(CO)OC1Oc2ccccc2N(=O)=O

InChI key

PXMQUEGJJUADKD-UHFFFAOYSA-N

form

solid

storage temp.

−20°C

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service