Skip to Content
MilliporeSigma

N5638

α-Naphtholbenzein

Synonym(s):

4,4′-(α-Hydroxybenzylidene)di-1-naphthol, p-naphtholbenzein

Sign Into View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C27H18O2
CAS Number:
Molecular Weight:
374.43
Beilstein/REAXYS Number:
2628063
UNSPSC Code:
12352103

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

pH

9.8-11.0, yellow to blue(Visual transition)

mp

230-235 °C (lit.)

SMILES string

Oc1ccc(\C(c2ccccc2)=C3\C=CC(=O)c4ccccc34)c5ccccc15

InChI

1S/C27H18O2/c28-25-16-14-23(19-10-4-6-12-21(19)25)27(18-8-2-1-3-9-18)24-15-17-26(29)22-13-7-5-11-20(22)24/h1-17,28H/b27-24-

InChI key

VDDWRTZCUJCDJM-PNHLSOANSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service