NEUR1050

Sigma-Aldrich

Neuronal Medium - 50mL

sterility

sterile

form

liquid

application(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Related Categories

Components

Supplement A
Supplement B
Supplement C

RIDADR

NONH for all modes of transport

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.