MilliporeSigma
All Photos(1)

O2505

Sigma-Aldrich

Palmityl oleate

~99%

Synonym(s):
cis-9-Octadecenoic acid hexadecanyl ester, Oleic acid palmityl ester
Empirical Formula (Hill Notation):
C34H66O2
CAS Number:
Molecular Weight:
506.89
EC Number:
MDL number:
PubChem Substance ID:

assay

~99%

functional group

ester
oleic acid

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCOC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C34H66O2/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34(35)36-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2/h17,19H,3-16,18,20-33H2,1-2H3/b19-17-

InChI key

JYTMDBGMUIAIQH-ZPHPHTNESA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service