Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O8126

Sigma-Aldrich

Orange II sodium salt

Dye content ~85 %

Synonym(s):

4-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid sodium salt, Acid Orange 7, Acid Orange A, Orange II, Tropaeolin 000 No. 2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N2NaO4S
CAS Number:
Molecular Weight:
350.32
Colour Index Number:
15510
Beilstein/REAXYS Number:
3898201
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ~85%

SMILES string

[Na+].Oc1ccc2ccccc2c1\N=N\c3ccc(cc3)S([O-])(=O)=O

InChI

1S/C16H12N2O4S.Na/c19-15-10-5-11-3-1-2-4-14(11)16(15)18-17-12-6-8-13(9-7-12)23(20,21)22;/h1-10,19H,(H,20,21,22);/q;+1/p-1/b18-17+;

InChI key

CQPFMGBJSMSXLP-ZAGWXBKKSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfei Ji et al.
Environmental technology, 33(4-6), 467-472 (2012-05-29)
CeO2 nanoparticles were prepared by a simple precipitation method, followed by calcination treatment. By selecting photocatalytic degradation of acid orange 7 (AO7) as a probe reaction, the influences of calcination temperature of catalyst, the concentration of AO7, initial pH value
Deepika Parasuraman et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(10), 2507-2515 (2012-04-28)
We review our recent efforts utilizing poly(N-isopropylacrylamide)-co-acrylic acid (pNIPAm-co-AAc) microgels and their assemblies for the removal of an azo-dye molecule, 4-(2-Hyrodxy-1-napthylazo) benzenesulfonic acid sodium salt (Orange II), from aqueous solutions. First, the ability of dispersed, single microgels to remove Orange
Soon-An Ong et al.
Journal of environmental sciences (China), 24(2), 291-296 (2012-06-05)
We investigated the decolorization of Orange II with and without the addition of co-substrates and nutrients under an anaerobic sequencing batch reactor (ASBR). The increase in COD concentrations from 900 to 1750 to 3730 mg/L in the system treating 100
Ashish Yadav et al.
Toxicology letters, 208(3), 239-245 (2011-12-06)
Orange II, an azo dye, is not permitted in food preparations, but high levels of the dye have been detected in different food commodities. Though there are reports on the toxicity of Orange II but knowledge based on the immunomodulatory
Yan-Qing Wang et al.
Journal of photochemistry and photobiology. B, Biology, 113, 14-21 (2012-05-30)
The interactions between human hemoglobin and orange I (or orange II) were investigated by UV/vis absorption, circular dichroism, fluorescence spectra techniques, and molecular modeling method. Orange I and orange II effectively quenched the intrinsic fluorescence of human hemoglobin by static

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service