MilliporeSigma
All Photos(1)

Documents

OH30057494

Ohaus® pH Meter

Model ST2100-B, AC/DC input 120 V, US 3-pin plug

Sign Into View Organizational & Contract Pricing

material

epoxy shaft

feature

CSA approved

packaging

pack of 1 ea

manufacturer/tradename

Ohaus® 30057494

ORP measuring range

-1999-+1999 mV

pH measuring range

-2-16 pH unit

temperature measuring range

0-100 °C

Looking for similar products? Visit Product Comparison Guide

General description

Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
The built-in electrode holder provides uncomplicated and convenient all-in-one operation while the large LCD display screen clearly displays results.

Application

pH, oxidation-reduction potential (ORP) measurement

Features and Benefits

Auto buffer recognition, two point calibration, quick reference guide
Automatic and manual temperature compensation, auto buffer recognition, calibration icon

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service