Skip to Content
MilliporeSigma

P0041

PF-750

≥98% (HPLC)

Synonym(s):

N-Phenyl-4-(quinolin-3-ylmethyl)piperidine-1-carboxamide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H23N3O
CAS Number:
Molecular Weight:
345.44
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

PF-750, ≥98% (HPLC)

InChI

1S/C22H23N3O/c26-22(24-20-7-2-1-3-8-20)25-12-10-17(11-13-25)14-18-15-19-6-4-5-9-21(19)23-16-18/h1-9,15-17H,10-14H2,(H,24,26)

SMILES string

O=C(Nc1ccccc1)N2CCC(CC2)Cc3cnc4ccccc4c3

InChI key

BIODYGOZWZNCAG-UHFFFAOYSA-N

assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

Biochem/physiol Actions

PF-750 is a potent, time-dependent, irreversible FAAH inhibitor.
PF-750 is a potent, time-dependent, irreversible FAAH inhibitor with IC50 values 0.6 and 0.016 μM when preincubated with recombinant human FAAH for 5 and 60 minutes, respectively. Fatty acid amide hydrolase (FAAH) is the enzyme responsible for hydrolysis and inactivation of fatty acid amides including anandamide and oleamide. Activity-based profiling of various human and murine tissue proteome samples revealed that PF-750 is highly selective for FAAH relative to other serine hydrolases, showing no discernable off-site activity up to 500 μM. PF-750 shows 10-fold better potency than URB597 (Sigma# U4133) after 30 min preincubation. PF-750 is highly selective on FAAH. Even at as high as 500 μM, it had no interactions with many tested enzymes, but URB597 and other known FAAH inhibitors did not perform well at low concentraction (100 μM).

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to cannabinoid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service