Skip to Content
MilliporeSigma

Skip To

P123

Phencyclidine hydrochloride

Synonym(s):

1-(1-Phenylcyclohexyl)piperidine hydrochloride, PCP hydrochloride

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C17H25N · HCl
CAS Number:
Molecular Weight:
279.85
UNSPSC Code:
12352200
MDL number:


drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada; Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

SMILES string

Cl.C1CCN(CC1)C2(CCCCC2)c3ccccc3

InChI

1S/C17H25N.ClH/c1-4-10-16(11-5-1)17(12-6-2-7-13-17)18-14-8-3-9-15-18;/h1,4-5,10-11H,2-3,6-9,12-15H2;1H

InChI key

BUAJNGPDPGKBGV-UHFFFAOYSA-N

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


Still not finding the right product?

Explore all of our products under Phencyclidine hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Philip L R Gaskin et al.
Psychopharmacology, 231(12), 2533-2545 (2014-01-10)
Schizophrenia is a debilitating disorder comprising positive, negative and cognitive deficits with a poorly defined neurobiological aetiology; therefore, animal models with greater translational reliability are essential to develop improved therapies. This study combines two developmental challenges in rats, neonatal phencyclidine
Bita Moghaddam et al.
Schizophrenia bulletin, 38(5), 942-949 (2012-08-18)
Here, we describe our collaborative efforts to use N-methyl-d-aspartate (NMDA) receptor antagonists as a translational tool to advance our understanding of the pathophysiology of schizophrenia and identify potential new targets for treatment of schizophrenia. We began these efforts in the
L Wei et al.
Osteoporosis international : a journal established as result of cooperation between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA, 25(8), 2089-2096 (2014-05-09)
Recently, the use of the pharmacological agent strontium ranelate has come to prominence for the treatment of osteoporosis. While much investigation is focused on preventing disease progression, here we fabricate strontium-containing scaffolds and show that they enhance bone defect healing