P2780

Sigma-Aldrich

Parathyroid Hormone Fragment 13-34 human

≥95% (HPLC)

Empirical Formula (Hill Notation):
C125H199N39O33S
CAS Number:
Molecular Weight:
2808.22
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC@H(NC(=O)C@H(CO)NC(=O)C@H(CC(N)=O)NC(=O)C@H(CC(C)C)NC(=O)C@H(Cc1cnHcn1)NC(=O)C@@H(N)CCCCN)C(=O)NC@@H(CCC(O)=O)C(=O)NC@@H(CCCNC(N)=N)C(=O)NC@@H(C(C)C)C(=O)NC@@H(CCC(O)=O)C(=O)NC@@H(Cc2cnHc3ccccc23)C(=O)NC@@H(CC(C)C)C(=O)NC@@H(CCCNC(N)=N)C(=O)NC@@H(CCCCN)C(=O)NC@@H(CCCCN)C(=O)NC@@H(CC(C)C)C(=O)NC@@H(CCC(N)=O)C(=O)NC@@H(CC(O)=O)C(=O)NC@@H(C(C)C)C(=O)NC@@H(Cc4cnHcn4)C(=O)NC@@H(CC(N)=O)C(=O)NC@@H(Cc5ccccc5)C(O)=O

InChI

1S/C125H199N39O33S/c1-63(2)47-83(112(185)149-79(34-37-94(130)166)107(180)159-91(56-99(173)174)119(192)164-101(67(9)10)122(195)160-88(53-71-59-138-62-143-71)116(189)158-89(54-95(131)167)117(190)161-92(123(196)197)50-68-25-13-12-14-26-68)153-105(178)76(31-19-22-43-128)145-103(176)75(30-18-21-42-127)144-104(177)77(32-23-44-139-124(133)134)147-111(184)84(48-64(3)4)154-114(187)86(51-69-57-141-74-29-16-15-27-72(69)74)156-108(181)81(36-39-98(171)172)151-121(194)100(66(7)8)163-110(183)78(33-24-45-140-125(135)136)146-106(179)80(35-38-97(169)170)148-109(182)82(40-46-198-11)150-120(193)93(60-165)162-118(191)90(55-96(132)168)157-113(186)85(49-65(5)6)155-115(188)87(52-70-58-137-61-142-70)152-102(175)73(129)28-17-20-41-126/h12-16,25-27,29,57-59,61-67,73,75-93,100-101,141,165H,17-24,28,30-56,60,126-129H2,1-11H3,(H2,130,166)(H2,131,167)(H2,132,168)(H,137,142)(H,138,143)(H,144,177)(H,145,176)(H,146,179)(H,147,184)(H,148,182)(H,149,185)(H,150,193)(H,151,194)(H,152,175)(H,153,178)(H,154,187)(H,155,188)(H,156,181)(H,157,186)(H,158,189)(H,159,180)(H,160,195)(H,161,190)(H,162,191)(H,163,183)(H,164,192)(H,169,170)(H,171,172)(H,173,174)(H,196,197)(H4,133,134,139)(H4,135,136,140)/t73-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,100-,101-/m0/s1

InChI key

VFUXFEHDZJLJEA-HQIUGSBTSA-N

Gene Information

human ... PTH(5741)

Related Categories

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Toshio Arakawa et al.
Clinical journal of the American Society of Nephrology : CJASN, 1(3), 525-531 (2007-08-21)
Measurement of bioactive parathyroid hormone (PTH) is essential for optimal management of bone abnormalities in dialysis patients. This can be accomplished by PTH measurements using third-generation PTH assays, which detect more or less of the first six amino acids of...
Ping Gao et al.
Clinical laboratory, 51(1-2), 21-29 (2005-02-22)
Most of what we know on PTH bioactivity has been associated with the first 34 amino acids of the PTH structure acting on the type I PTH/PTHrP receptor, leaving little place to the carboxyl-terminal structure. This reality has dictated the...
D Goltzman et al.
The Journal of biological chemistry, 250(8), 3199-3203 (1975-04-25)
Two synthetic analogues of bovine parathyroid hormone (PTH) with NH2-terminal modifications, PTH-(3-34) and [desamino-Ala-1]PTH-(1-34), were found to lack agonist activity but to demonstrate antagonist properties when tested in the rat renal cortical adenylyl cyclase assay in vitro against the native...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.