Documents

P7384

Sigma-Aldrich

Propyl palmitate

≥99%

Synonym(s):
Palmitic acid propyl ester
Empirical Formula (Hill Notation):
C19H38O2
CAS Number:
Molecular Weight:
298.50
EC Number:
MDL number:

assay

≥99%

density

0.85 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCCC

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service