Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P9607

Sigma-Aldrich

Anti-Potassium Channel Kv2.1 (DRK1) antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing

Select a Size

0.05 ML
$337.00

$337.00


Available to ship onApril 15, 2025Details



Select a Size

Change View
0.05 ML
$337.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$337.00


Available to ship onApril 15, 2025Details


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot (chemiluminescent): 1:250-1:500

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNB1(3745)
mouse ... Kcnb1(16500)
rat ... Kcnb1(25736)

Immunogen

synthetic peptide corresponding to amino acids 837-853 of rat potassium channel Kv2.1 (with an additional N-terminal cysteine and tyrosine).The epitope is identical in mouse and rat and highly homologous in human and pig.

Physical form

Lyophilized at ∼1 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.05% sodium azide.

Preparation Note

Protein A purified.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Maiya et al.
Genes, brain, and behavior, 6(1), 97-106 (2006-04-29)
Monoamine transporters play a key role in neuronal signaling by mediating reuptake of neurotransmitters from the synapse. The function of the dopamine transporter (DAT), an important member of this family of transporters, is regulated by multiple signaling mechanisms, which result

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service