Documents

PROBE3

Sigma-Aldrich

SulfoProbe DNA detection system

suitable for nucleic acid hybridization

usage

 kit sufficient for 1 mg of DNA probe lableing and detection (1000 cm2 of membrane.)

suitability

suitable for nucleic acid hybridization

storage temp.

2-8°C

Application

The SulfoProbe kit is a non-radioactive system for detecting chemically modified DNA probes hybridized to specific DNAs immobilized on nylon or nitrocellulose membranes.

Features and Benefits

  • Probe modification and detection reagents are included in a single kit.
  • DNA probe preparation is a simple chemical reaction using non-carcinogenic reagents.
  • No post-modification purification is necessary
  • Probes are stable for several months at −20°C.
  • Non-radioactive detection system.

Kit Components Also Available Separately

Product No.
Description
SDS

  • A3562Anti-Mouse IgG Alkaline Phosphatase Conjugate .08 mL

  • B56555-Bromo-4-chloro-3-indolyl phosphate BCIP)/Nitro Blue Tetrazolium NBT) 20 tablets

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.