PRS2509

Sigma-Aldrich

Anti-Caspase-14 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-Caspase 14, Anti-Mini-ICE, Anti-Mini-interleukin-1 converting enzyme, Anti-MICE
MDL number:
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 28 kDa

species reactivity

rat, mouse, human

application(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Casp14(12365)

Immunogen

a 16 amino acid peptide from near the carboxy-terminus of human caspase-14.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2509.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service