QPREST30020

Sigma-Aldrich

SILuPrEST CHSTF

SILuPrESTs Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase, B-cell RAG-associated gene protein
Pricing and availability is not currently available.

recombinant

expressed in E. coli LysA ArgA BL21(DE3)

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 34kDa including tags

purified by

immobilized metal affinity chromatography (IMAC)

packaging

pkg of 1nmol × 5 vials

storage condition

avoid repeated freeze/thaw cycles

concentration

(Lot specific concentration determined by ESI-MS/MS using highly pure and amino acid analyzed His6ABP as internal standard for targeted quantification of the labeled His6ABP-containing SILuPrEST will be stated on the Certificate of Analysis)

immunogen sequence

INSITTRIEFTTRQLPDLEDLKKQELHMFSVIPNKFLPNSKSPCWYEEFSGQNTTDPYLTNSYVLYSKRFRSTFDALRKAFWGHLAHAHGKHFRLRCLPHFYIIGQPKCGTTDLYDRLRLHPEVKFSAIKEPHWWTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CHSTF(51363)

General description

Lys, Arg 13C and 15N metabolically labeled recombinant human protein fragment

Application

Internal standard in MS-based quantitative proteomics

Physical form

Heavy proteins are provided in solution of 1M Urea PBS, pH 7.4

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Analysis Note

A purification and quantification tag (QTag) consisting of a hexahistidine (His6) sequence followed by an Albumin Binding Protein (ABP) domain derived from Streptococcal Protein G.
All SILuPrESTs contain an N-terminal His6ABP fusion tag, used for accurate determination of concentration. The fusion tag sequence is:

GSSHHHHHHSSGLVPRGSHMASLAEAKVLANRELDKYGVSDYHKNLINNAKTVEGVKDLQAQVVESAKKARISEATDGLSDFLKSQTPAEDTVKSIELAEAKVLANRELDKYGVSDYYKNLINNAKTVEGVKALIDEILAALPGTFAHYMDPNSSSVDKLAAA.

The specific human protein sequence begins directly after ′VDKLAAA′.
Isotopic Label Incorporation

The SILuPrEST standards are produced using metabolic labeling with heavy isotope labeled (15N, 13C) Lysine and Arginine residues, with greater than 99% isotopic label incorporation.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com
SILu is a trademark of Sigma-Aldrich Co. LLC

RIDADR

NONH for all modes of transport

WGK Germany

1

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.