Skip to Content
MilliporeSigma

R2756

EcoR V from Escherichia coli

buffered aqueous glycerol solution

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
UNSPSC Code:
12352204
MDL number:
EC Number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

Molecular Biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Application

EcoRV is a DNA restriction endonuclease used in molecular biology applications to cleave the recognition site 5′-GAT/ATC-3′, generating DNA fragments with blunt termini.

Biochem/physiol Actions

Recognition sequence: 5′-GAT/ATC-3′
Ligation and recutting results: After 2-10-fold Eco RV overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >85% of fragments can be ligated, and >95% recut.
Heat inactivation: 80 °C for 20 minutes.

Physical form

Solution in 20 mM Tris-HCl, pH 7.5, 1 mM EDTA, 0.1 mM PMSF, 100 mM NaCl, 10 mM 2-mercaptoethanol, 0.02% polydocanol (v/v), 0.1 mM PMSF, 50% glycerol (v/v) at 4°C

Other Notes

Supplied with 10x Restriction Endonuclease Buffer SB (B8781).

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Schildkraut et al.
Gene, 27(3), 327-329 (1984-03-01)
The cleavage site for the restriction endonuclease EcoRV has been found to be 5'-GAT/ATC-3', rather than 5'- GATAT /C-3' as reported earlier by Kholmina et al. [ Dokl . Akad . Nauk . 253 (1980) 495-497].
Stability of transformed antagonistic Fusarium oxysporum strains in vitro and in soil microcosms.
Migheli, Q., et al.
Molecular Ecology, 5, 641-641 (1996)
Saqib H Ansari et al.
Journal of pediatric hematology/oncology, 35(4), e153-e156 (2013-02-08)
β-thalassemia is characterized by impaired β-chain synthesis leading to ineffective erythropoiesis, severe anemia, and a need for blood transfusion. Presence of Xmn I polymorphism (-158 C-T nucleotide change) in γ-globin gene is associated with a higher fetal hemoglobin and a
Jaroslav Jelinek et al.
Epigenetics, 7(12), 1368-1378 (2012-10-19)
Genome wide analysis of DNA methylation provides important information in a variety of diseases, including cancer. Here, we describe a simple method, Digital Restriction Enzyme Analysis of Methylation (DREAM), based on next generation sequencing analysis of methylation-specific signatures created by
Amal M El-Shehaby et al.
Scandinavian journal of clinical and laboratory investigation, 73(1), 75-81 (2012-12-04)
Left ventricular hypertrophy (LVH) is a common manifestation of cardiovascular disease and has an important prognostic value in patients with end-stage renal disease (ESRD). Vitamin D receptor (VDR) has been intensively investigated, and one of these (BsmI) already has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service