MilliporeSigma
All Photos(1)

Documents

R4507

Sigma-Aldrich

Rose Bengal sodium salt

certified by the Biological Stain Commission

Sign Into View Organizational & Contract Pricing

Synonym(s):
4,5,6,7-Tetrachloro-2′,4′,5′,7′-tetraiodofluorescein sodium salt, Acid Red 94, Bengal Rose B sodium salt
Empirical Formula (Hill Notation):
C20H2Cl4I4Na2O5
CAS Number:
Molecular Weight:
1017.64
Colour Index Number:
45440
Beilstein:
3645857
EC Number:
MDL number:
PubChem Substance ID:

grade

certified by the Biological Stain Commission

composition

Dye content, ≥80%

SMILES string

[Na+].[Na+].[O-]C(=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1C2=C3C=C(I)C(=O)C(I)=C3Oc4c(I)c([O-])c(I)cc24

InChI

1S/C20H4Cl4I4O5.2Na/c21-10-8(9(20(31)32)11(22)13(24)12(10)23)7-3-1-5(25)16(29)14(27)18(3)33-19-4(7)2-6(26)17(30)15(19)28;;/h1-2,29H,(H,31,32);;/q;2*+1/p-2

InChI key

UWBXIFCTIZXXLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in Conn′s technique for bacteria in soil; as an alternative to phloxine B in Kreyberg′s stain for keratin and mucus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

S S Pashin et al.
Bulletin of experimental biology and medicine, 168(4), 525-528 (2020-03-18)
Typical ischemic damage to neurons were detected in the focus of experimental photothrombosis and in the transition zone. They were associated with symptoms of impaired motor functions and dysfunction of pelvic organs. The applied method of focal photothrombosis can be
D W Marshall et al.
Appetite, 39(3), 193-206 (2002-12-24)
Most people get married or spend time living with a partner at some stage in their lives. But what effect does this change have, if any, on their eating habits? The transition from bachelor or spinster to young couple represents
Iu D Shenin et al.
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic], 41(2), 16-21 (1996-02-01)
The study was aimed at the development of a laboratory procedure for isomerization of polyene macrolides in the presence of sensitizers. The reaction was performed with exposure of levorin solutions to the xenon lamp irradiation in the presence of rose
A Ito et al.
Journal of the National Cancer Institute, 77(1), 277-281 (1986-07-01)
The tumorigenicity of 9-3',4',5',6'-tetrachloro-o-carboxy phenyl-6-hydroxy-2,4,5,7-tetraiodo-3-isoxanthone sodium (CAS: 632-68-8) [also called Food Red 105 (FR 105) or Rose Bengal B], which is widely used for food or cosmetic coloring in Japan, was examined in (C57BL/6N X C3H/N)F1 mice. Animals were divided
Y Hiasa et al.
Japanese journal of cancer research : Gann, 79(3), 314-319 (1988-03-01)
The effects of erythrosine (Red 3), rose bengal B (Red 105) and thyroidectomy on the development of thyroid tumor were examined in male Wistar rats treated with N-bis(2-hydroxypropyl)nitrosamine (DHPN). Red 3 and Red 105 were used at 4% in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service