MilliporeSigma
All Photos(1)

R5523

Sigma-Aldrich

REV 5901

Synonym(s):
α-Pentyl-3-[2-quinolinylmethoxy]benzyl alcohol
Empirical Formula (Hill Notation):
C22H25NO2
CAS Number:
Molecular Weight:
335.44
MDL number:
PubChem Substance ID:

form

solid

SMILES string

CCCCCC(O)c1cccc(OCc2ccc3ccccc3n2)c1

InChI

1S/C22H25NO2/c1-2-3-4-12-22(24)18-9-7-10-20(15-18)25-16-19-14-13-17-8-5-6-11-21(17)23-19/h5-11,13-15,22,24H,2-4,12,16H2,1H3

InChI key

JRLOEMCOOZSCQP-UHFFFAOYSA-N

Gene Information

human ... DRD4(1815)
rat ... Alox5(25290)

Biochem/physiol Actions

Leukotriene D4 receptor antagonist.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service