R882N

Sigma-Aldrich

Rat Cortical Neurons: RCoN

NACRES:
NA.81

Quality Level

biological source

rat brain (cortex)

packaging

pkg of 2,000,000 cells (R882N-20)
pkg of 4,000,000 cells (R882N-40)

growth mode

Adherent

morphology

Adherent. When cultured under the recommended conditions, cells arborize and form complex neurite network in one week.

application(s)

cell culture | mammalian: suitable

storage temp.

−196°C

Related Categories

General description

Rat cortical neurons (RCoN) are derived from cerebral cortices of day 18 embryonic Sprague Dawley rat brains. Cryopreserved immediately after isolation.

Cell Line Origin

Brain

Application

Basic research on cognition, memory, attention, perceptual awareness, thought, language, consciousness, signal transmission, body control and regulation of the other parts of the body, brain dysfunction and degeneration, brain injuries, neurological and psychiatric disorders, disease mechanisms, pathophysiologies, disease mechanisms, pathophysiologies, drug screening and development, toxicity testing, immunostaining, live cell imaging, co-culturing, electrophysiology.

Packaging

R882N-40 4,000,000 cells in Serum-free Freezing Medium
R882N-20 2,000,000 cells in Serum-free Freezing Medium

Components

Serum-free Freezing Medium

Quality

Each lot was tested for proper morphology, Stain positive for β III-Tubulin, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Certificate of Analysis
Certificate of Origin
Articles
Products to enable the creation of neural models specific for your research.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.