Documents

RDC1

Sigma-Aldrich

Director Universal PCR System

Pricing and availability is not currently available.

usage

sufficient for 25 PCR reactions

shipped in

wet ice

storage temp.

−20°C

Related Categories

Application

The simple three step procedure (PCR, Exonuclease III digestion and rapid ligation/transformation) can be completed in one day. The typical cloning efficiency using this method is greater than 80%.

Features and Benefits

  • Universal - PCR amplicon can be cloned into any expression vector
  • High Cloning Efficiency - Typically >80%
  • High Expression Efficiency - Typically >66%
  • High Fidelity - Long and accurate, hotstart enzyme generates amplicons up to 20 kb with fidelity up to 6.5× greater than standard Taq DNA polymerase
  • Fast - Simple three-step procedure allows completion in less than one day

Other Notes

The Director Universal Cloning System provides a simple, rapid and universal method to directionally clone PCR products into a vector cleaved with 5′ overhang-producing restriction endonucleases.

Principle

Directionality is achieved by pairing directionally designed PCR primers (e.g., containing restriction sites) with any appropriately digested plasmid. The kit contains an optimized nucleotide triphosphate mix, containing dATPαS and dGTPαS, that is used for the PCR step. After PCR, the cohesive 5′ termini of the amplicon are generated by Exonuclease III digestion instead of being generated by traditional restriction enzyme digestions. The dA/GTPαS that was incorporated into the amplicon during PCR protects it from over-digestion by Exonuclease III. The nucleotide mix in the kit is specially formulated so that the amplicon terminates at a statistically determined array of 3′ dA/GαS sites. PCR primers are designed such that the 5′ termini complement the 5′ overhangs of the predigested plasmid.

Legal Information

Director is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • 10x AccuTaq™ LA DNA Polymerase Buffer 500 μL

  • Control PCR Template, 1ng/μl 10 μL

  • Control RDC primer-R (with 5′ phosphorylation) 25 μL

  • Control RDC primer-F (with 5′ phosphorylation) 25 μL

  • ExoNuclease III, 100 units/μl 25 μL

  • JumpStart™ REDAccuTaq™ LA DNA Polymerase, 1 unit/μl 62.5 μL

  • Molecular biology grade water 500 μL

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.