MilliporeSigma
All Photos(1)

Documents

S2752

Sigma-Aldrich

Tin(II) chloride

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing

Synonym(s):
Stannous chloride
Linear Formula:
SnCl2
CAS Number:
Molecular Weight:
189.62
EC Number:
MDL number:
PubChem Substance ID:

product line

ReagentPlus®

Assay

≥99%

bp

652 °C (lit.)

mp

246 °C (lit.)

SMILES string

Cl[SnH2]Cl

InChI

1S/2ClH.Sn/h2*1H;/q;;+2/p-2

InChI key

AXZWODMDQAVCJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Wei-Hsin Hsu et al.
Nuclear medicine and biology, 41(9), 765-771 (2014-07-17)
Liposome in delivering radionuclide for cancer therapy has been expansively studied; however, liposome itself can be deliberately entrapped and destroyed by the reticuloendothelial system, causing an insufficiency of the drug delivery, which in turn would restrict the effectiveness of the
Shi Gao et al.
Clinical nuclear medicine, 39(1), e14-e19 (2013-08-07)
Tc N-MPO ([Tc N(MPO)(PNP5)]: HMPO = 2-mercaptopyridine N-oxide, and PNP5 = N-ethoxyethyl-N,N-bis[2-(bis(3-methoxypropyl)phosphino)ethyl]amine) is a new Tc radiotracer useful for myocardial perfusion imaging. This study was designed to determine its pharmacokinetics and biodistribution in healthy volunteers. Ten healthy volunteers were involved
Sebastiaan Wesseling et al.
British journal of pharmacology, 172(6), 1607-1619 (2014-03-07)
Whether NO, carbon monoxide (CO) and hydrogen sulfide (H2 S) compensate for each other when one or more is depleted is unclear. Inhibiting NOS causes hypertension and kidney injury. Both global depletion of H2 S by cystathionine γ-lyase (CSE) gene
Lars Bellner et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(1), 105-115 (2014-10-25)
Heme oxygenase (HO)-2 deficiency impairs wound healing and exacerbates inflammation following injury. We examine the impact of HO-2 deficiency on macrophage function and the contribution of macrophage HO-2 to inflammatory and repair responses to injury. Corneal epithelial debridement was performed
Rajendran Suresh et al.
The Journal of organic chemistry, 77(3), 1468-1476 (2012-01-11)
The synthesis of 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)quinolines by a SnCl(2)-catalyzed multicomponent reaction has been described. The reaction proceeds chemo- and regioselectively in an atom-economic way, generating a library of 24 quinoline derivatives.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service