S3938

Sigma-Aldrich

SeqSaver Sequencing Premix Dilution Buffer

for DNA sequencing

NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

grade

for molecular biology

form

liquid

storage temp.

−20°C

Application

Suitable for use with the ABI BigDye® terminator premix solution for sequencing.

Features and Benefits

  • Reduces the amount of Terminator Premix required
  • Compatable with BigDye®, DYEnamic ETs, ThermoSequemase II
  • No reduction in resolution, read length, or signal strength
  • Compatable with ABI Prism 3700, 3100 & 310 DNA sequencers
  • Also compatable with Molecular Dynamics MegaBACE instruments

Legal Information

BigDye is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
SeqSaver is a trademark of Sigma-Aldrich Co. LLC

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Identification of SNPs, or Mutations in Sequence Chromatograms
Draper N
Meths. Mol. Biol., 439, 35-52 (2008)
L Wen
Molecular biotechnology, 17(2), 135-142 (2001-06-09)
The use of automated fluorescent DNA sequencer systems and PCR-based DNA sequencing methods plays an important role in the actual effort to improve the efficiency of large-scale DNA analysis. While dideoxy-terminators labeled with energy-transfer dyes (BigDyes) provide the most versatile...
Anthony J Aldave et al.
Investigative ophthalmology & visual science, 51(8), 4006-4012 (2010-04-02)
To identify the genetic basis of posterior amorphous corneal dystrophy (PACD) segregating in a large pedigree. The authors performed clinical evaluation of a previously unreported pedigree with PACD, light and electron microscopic examination of an excised corneal button, genomewide linkage...
Vivek S Yellore et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 9(4), 228-234 (2007-04-18)
The study purpose was to identify the genetic basis of posterior polymorphous corneal dystrophy, an autosomal dominant disorder of the corneal endothelium that is associated with the development of corneal edema, necessitating corneal transplantation for visual rehabilitation. Glaucoma also develops...
Anthony J Aldave et al.
American journal of medical genetics. Part A, 143A(21), 2549-2556 (2007-10-16)
Mutations in the two-handed zinc-finger homeodomain transcription factor gene (TCF8) have been associated with posterior polymorphous corneal dystrophy (PPCD) and extraocular developmental abnormalities. We performed screening of TCF8 in 32 affected, unrelated probands, affected and unaffected family members of probands...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.