Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C24H26N2O6
CAS Number:
Molecular Weight:
438.47
UNSPSC Code:
12352202
PubChem Substance ID:
EC Number:
248-477-6
MDL number:
form
solid
SMILES string
CCCCC1(COC(=O)CCC(O)=O)C(=O)N(N(C1=O)c2ccccc2)c3ccccc3
InChI
1S/C24H26N2O6/c1-2-3-16-24(17-32-21(29)15-14-20(27)28)22(30)25(18-10-6-4-7-11-18)26(23(24)31)19-12-8-5-9-13-19/h4-13H,2-3,14-17H2,1H3,(H,27,28)
InChI key
ONWXNHPOAGOMTG-UHFFFAOYSA-N
Biochem/physiol Actions
A prodrug of phenylbutazone.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[The biological fate of 4-butyl-4-(beta-carboxypropionyloxymethyl)-1,2-diphenyl-3,5-pyrazolidinedione(suxibuzone), antiinflammatory drug. I. Plasma levels and urinary excretion after oral administration of suxibuzone (author's transl)].
T Shindo et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 99(12), 1186-1200 (1979-12-01)
[The biological fate of 4-butyl-4-(beta-carboxypropionyloxymethyl)-1,2-diphenyl-3,5-pyrazolidinedione (suxibuzone), antiinflammatory drug. II. The biological fate of suxibuzone (SB) after daily oral administration (author's transl)].
K Taira et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 100(3), 272-279 (1980-03-01)
S Yanagi et al.
Cancer letters, 36(1), 11-18 (1987-07-01)
Among phenylbutazone (PZ) and its related compounds, suxibuzone (SUX) caused the most extensive decrease in pyruvate kinase (PK) activity with lower toxicity. Therefore, we studied the effect of SUX on rat hepatocarcinogenesis to confirm our assumption that an agent which
L Monreal et al.
Research in veterinary science, 76(2), 145-149 (2003-12-16)
The objective was to compare the gastrointestinal and general toxicity of suxibuzone (SBZ) to that of phenylbutazone (PBZ) when administered orally in horses. Fifteen healthy horses were allocated to three treatment groups. One group received a high dose of PBZ
Frank M Andrews et al.
Veterinary therapeutics : research in applied veterinary medicine, 10(3), 113-120 (2009-12-30)
Eighteen mature, healthy horses were divided into three groups (six per group) receiving either no treatment, 15 consecutive days of phenylbutazone (PBZ), or 15 consecutive days of suxibuzone (SBZ) at recommended label doses. Horses underwent endoscopy before and after the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service