Skip to Content
MilliporeSigma

S6566

Sodium phosphate monobasic

Biotechnology Performance Certified, suitable for insect cell culture

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
PubChem Substance ID:
eCl@ss:
38070208
UNSPSC Code:
12161700
EC Number:
231-449-2
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Sodium phosphate monobasic, Biotechnology Performance Certified, suitable for insect cell culture

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

SMILES string

[Na+].OP(O)([O-])=O

grade

Biotechnology Performance Certified

form

powder

technique(s)

cell culture | insect: suitable

impurities

Bioburden, tested
endotoxin, tested
≤1% water

pH

4.0-4.5 (25 °C, 50 g/L in water)

anion traces

chloride (Cl-): ≤40 ppm

cation traces

Fe: ≤30 ppm
heavy metals (as Pb): ≤10 ppm

absorption

≤0.2 at 260 in H2O at 1 M
≤0.2 at 280 in H2O at 1 M

application(s)

diagnostic assay manufacturing

foreign activity

DNase, RNase, Protease, none detected

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Adler et al.
Gastrointestinal endoscopy, 76(6), 1170-1174 (2012-10-03)
Colon capsule endoscopy (CCE) is a noninvasive technique for the detection of colorectal lesions. However, for CCE to be offered as an out-of-clinic procedure, the system needs to automatically alert the patient when to ingest the laxative (booster). We tested
Charles W Kimbrough et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(20), 4576-4585 (2015-07-01)
pH-low insertion peptides (pHLIP) can serve as a targeting moiety that enables pH-sensitive probes to detect solid tumors. Using these probes in conjunction with multispectral optoacoustic tomography (MSOT) is a promising approach to improve imaging for pancreatic cancer. A pH-sensitive
Johanna Troberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 107-118 (2014-10-11)
Understanding drug glucuronidation in the dog, a preclinical animal, is important but currently poorly characterized at the level of individual enzymes. We have constructed cDNAs for the 10 dog UDP-glucuronosyltransferases of subfamily 1A (dUGT1As), expressed them in insect cells, and
Filis Kazazi-Hyseni et al.
Pharmaceutical research, 31(10), 2844-2856 (2014-05-16)
The aim of this study was the development of poly(D,L-lactide-co-glycolide) (PLGA) microspheres with controlled porosity, to obtain microspheres that afford continuous release of a macromolecular model compound (blue dextran). PLGA microspheres with a size of around 40 μm and narrow size
Sima Rahimian et al.
Biomaterials, 61, 33-40 (2015-05-21)
This study investigated the feasibility of the use of polymeric microparticles for sustained and local delivery of immunomodulatory antibodies in immunotherapy of cancer. Local delivery of potent immunomodulatory antibodies avoids unwanted systemic side effects while retaining their anti-tumor effects. Microparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service