S8388

Sigma-Aldrich

Sodium acetate solution

0.3 M

Empirical Formula (Hill Notation):
C2H3NaO2
CAS Number:
Molecular Weight:
82.03
Beilstein/REAXYS Number:
3595639
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

form

crystalline

concentration

0.3 M

pH

8.5-9.9 (25 °C, 246 g/L)

solubility

water: soluble 246 g/L at 20 °C

storage temp.

2-8°C

SMILES string

Na+.CC(O-)=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

> 250 °C - closed cup

Amanda P De Souza et al.
Journal of experimental botany, 66(14), 4351-4365 (2015-04-25)
The production of bioenergy from grasses has been developing quickly during the last decade, with Miscanthus being among the most important choices for production of bioethanol. However, one of the key barriers to producing bioethanol is the lack of information...
Carol L Ladner-Keay et al.
PloS one, 9(6), e98753-e98753 (2014-06-04)
The formation of β-sheet rich prion oligomers and fibrils from native prion protein (PrP) is thought to be a key step in the development of prion diseases. Many methods are available to convert recombinant prion protein into β-sheet rich fibrils...
S Srinivasa et al.
The British journal of surgery, 100(1), 66-74 (2012-11-08)
Goal-directed fluid therapy (GDFT) has been compared with liberal fluid administration in non-optimized perioperative settings. It is not known whether GDFT is of value within an enhanced recovery protocol incorporating fluid restriction. This study evaluated GDFT under these circumstances in...
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace...
Do Gyun Lee et al.
Water research, 46(13), 4226-4234 (2012-06-08)
Triclosan, a synthetic antimicrobial agent, has been considered as an emerging environmental contaminant. Here we reported a triclosan-degrading wastewater bacterial isolate, Sphingopyxis strain KCY1, capable of dechlorinating triclosan with a stoichiometric release of chloride. The stain can degrade diphenyl ether...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.