SAB1400834

Sigma-Aldrich

Anti-ZNF509 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):
Anti-MGC126280, Anti-FLJ38559, Anti-MGC126279
NACRES:
NA.41
Pricing and availability is not currently available.

Quality Level

biological source

mouse

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

application(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

conjugate

unconjugated

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ZNF509(166793)

Immunogen

ZNF509 (AAH16477.1, 1 a.a. ~ 248 a.a) full-length human protein.

Sequence
MQRKLVKHRIRHTGERPYSCSACGKCFGGSGDLRRHVRTHTGEKPYTCEICNKCFTRSAVLRRHKKMHCKAGDESPDVLEELSQAIETSDLEKSQSSDSFSQDTSVTLMPVSVKLPVHPVENSVVEFDSHSGGSYCKLRSMIQPHGVSDQEKLSLDPGKLAKPQMQQTQPQAYAYSDVDTPAGGEPLQADGMAMIRSSLAALDNHGGDPLGSRASSTTYRNSEGQFFSSMTLWGLAMKTLQNENELDQ

Physical form

Solution in phosphate buffered saline, pH 7.4

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.