Skip to Content
MilliporeSigma

SAB1405271

Monoclonal Anti-PRKRIP1 antibody produced in mouse

clone 4D11-3F11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

C114, FLJ13902, FLJ40957

Sign In to View Organizational & Contract Pricing.

Select a Size

100 μG

$580.00

$580.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D11-3F11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~46.35 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKRIP1(79706)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB1409817WH0023613M1SAB1404983
clone

4D11-3F11, monoclonal

clone

1F6-3C2, monoclonal

clone

5B12, monoclonal

clone

6A12, monoclonal

Gene Information

human ... PRKRIP1(79706)

Gene Information

human ... PRKRIP1(79706)

Gene Information

human ... ZMYND8(23613)

Gene Information

human ... ZMYND8(23613)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Immunogen

PRKRIP1 (AAH14298, 1 a.a. ~ 184 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MASPAASSVRPPRPKKEPQTLVIPKNAAEEQKLKLERLMKNPDKAVPIPEKMSEWAPRPPPEFVRDVMGSSAGAGSGEFHVYRHLRRREYQRQDYMDAMAEKQKLYAEFQKRLEKNKIAAEEQTAKRRKKRQKLKEKKLLAKKMKLEQKKQEGPGQPKEQGSSSSAEASGTEEEEEVPSFTMGR

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service