MilliporeSigma
  • SAB1408047
All Photos(1)

SAB1408047

Sigma-Aldrich

Anti-ITM2C antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):
BRI3, BRICD2C, E25, E25C, ITM3
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~25.41 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ITM2C(81618)

Immunogen

ITM2C (AAH25742, 1 a.a. ~ 230 a.a) full-length human protein.

Sequence
MVKISFQPAVAGIKGDKADKASASAPAPASATEILLTPAREEQPPQHRSKRGSSVGGVCYLSMGMVVLLMGLVFASVYIYRYFFLAQLARDNFFRCGVLYEDSLSSQVRTQMELEEDVKIYLDENYERINVPVPQFGGGDPADIIHDFQRRGTYLPQTYIIQEEMVVTEHVSDKEALGSFIYHLCNGKDTYRLRRRATRRRINKRGAKNCNAIRHFENTFVVETLICGVV

Packaging

50 μg

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service