SAB1408129

Sigma-Aldrich

Anti-ZNF503 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):
FLJ45745, MGC2555, NOLZ-1
NACRES:
NA.41
Pricing and availability is not currently available.

Quality Level

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~62.6 kDa

species reactivity

human

application(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ZNF503(84858)

Immunogen

ZNF503 (NP_116161, 1 a.a. ~ 646 a.a) full-length human protein.

Sequence
MSTAPSLSALRSSKHSGGGGGGGGGGGADPAWTSALSGNSSGPGPGSSPAGSTKPFVHAVPPSDPLRQANRLPIKVLKMLTARTGHILHPEYLQPLPSTPVSPIELDAKKSPLALLAQTCSQIGKPDPSPSSKLSSVASNGGGAGGAGGGAAGDKDTKSGPLKLSDIGVEDKSSFKPYSKPGSDKKEPGGGGGGGGGGGGGGGGVSSEKSGFRVPSATCQPFTPRTGSPSSSASACSPGGMLSSAGGAPEGKDDKKDTDVGGGGKGTGGASAEGGPTGLAHGRISCGGGINVDVNQHPDGGPGGKALGSDCGGSSGSSSGSGPSAPTSSSVLGSGLVAPVSPYKPGQTVFPLPPAGMTYPGSLAGAYAGYPPQFLPHGVALDPTKPGSLVGAQLAAAAAGSLGCSKPAGSSPLAGASPPSVMTASLCRDPYCLSYHCASHLAGAAAASASCAHDPAAAAAALKSGYPLVYPTHPLHGVHSSLTAAAAAGATPPSLAGHPLYPYGFMLPNDPLPHICNWVSANGPCDKRFATSEELLSHLRTHTAFPGTDKLLSGYPSSSSLASAAAAAMACHMHIPTSGAPGSPGTLALRSPHHALGLSSRYHPYSKSPLPTPGAPVPVPAATGPYYSPYALYGQRLTTASALGYQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.