SAB2500150

Sigma-Aldrich

Anti-BAP/SIL1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-BiP-associated protein

Quality Level

biological source

goat

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

application(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SIL1(64374)

Immunogen

Peptide with sequence C-ELLGSVNSLLKELR from the C Terminus of the protein sequence according to NP_071909.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

hazcat

Eye Irrit. 2 - Skin Irrit. 2

storage_class_code

12 - Non Combustible Liquids

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service