MilliporeSigma
  • SAB2500338
All Photos(1)

SAB2500338

Sigma-Aldrich

Anti-DYX1C1/EKN1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-DYX1, Anti-EKN1, Anti-FLJ37882
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, human

technique(s)

indirect ELISA: suitable
western blot: suitable

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... DYX1C1(161582)

Immunogen

Peptide with sequence PLQVSDYSWQQTKT-C from the N Terminus of the protein sequence according to NP_570722.2; NP_001028731.1; NP_001028732.1.

Packaging

100 μg

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service