SAB2500916

Sigma-Aldrich

Anti-SDHB antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-SDHIP, Anti-IP, Anti-SDH, Anti-SDH1
MDL number:
NACRES:
NA.41

Quality Level

biological source

goat

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

application(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SDHB(6390)

Immunogen

Peptide with sequence C-ATYKEKKASV from the C Terminus of the protein sequence according to NP_002991.1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Letizia Canu et al.
The Journal of clinical endocrinology and metabolism, 99(7), 2321-2326 (2014-04-25)
About 35% of patients with pheochromocytoma/paraganglioma carry a germline mutation in one of the 10 main susceptibility genes. The recent introduction of next-generation sequencing will allow the analysis of all these genes in one run. When positive, the analysis is...
Xiao-Hui Zou et al.
Virology, 468-470, 388-396 (2014-09-23)
The human adenovirus (HAdV) early protein E1B55K interacts with E4orf6 to form an E3 ubiquitin ligase complex, which plays key roles in virus replication. To illustrate the reason for the fastidiousness of HAdV-41 in 293 cells, interaction between heterotypic E1B55K...
Jingnan Sun et al.
Nucleic acids research, 42(15), 9588-9601 (2014-08-06)
Dysregulation of the insulin-like growth factor type I receptor (IGF1R) has been implicated in the progression and therapeutic resistance of malignancies. In acute myeloid leukemia (AML) cells, IGF1R is one of the most abundantly phosphorylated receptor tyrosine kinases, promoting cell...
Manuraj Pandey et al.
Toxicology and applied pharmacology, 280(2), 296-304 (2014-08-30)
In the present study, we showed the correlation of EZH2, SUV39H1 or G9a expression and histone modifications with the urethane induced mouse lung tumorigenesis in the presence or absence of antitumor agent, inositol hexaphosphate (IP6). Tumorigenesis and the molecular events...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service