SAB2701816

Sigma-Aldrich

Anti-ttll1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):
zgc:63993, ttll1, MGC63993
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

zebrafish

application(s)

immunohistochemistry: suitable
western blot: suitable

conjugate

unconjugated

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

zebrafish ... ttll1(393994)

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 24 and 179 of ttll1 according to NP_957313

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.