SAB2702009

Sigma-Aldrich

Anti-TRIM37 (C-terminal) antibody produced in rabbit

Synonym(s):
MUL, POB1, TEF3
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

108 kDa

species reactivity

human

concentration

1.02 mg/mL

application(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 500-3000

conjugate

unconjugated

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TRIM37(4591)

Immunogen

Synthetic peptide corresponding to a region within amino acids 902 and 964 of TRIM37 (Uniprot ID#O94972)

Application

Anti-TRIM37 (C-terminal) antibody produced in rabbit is suitable for immunocytochemistry, immunofluorescence, immunohistochemistry (formalin-fixed, paraffin-embedded sections) and western blot.

Biochem/physiol Actions

TRIM37 (Tripartite motif containing 37) restricts centriole reduplication during centriole biogenesis. It participates in epigenetic transcriptional repression by facilitating monoubiquitination of ′Lys-119′ of histone H2A (H2AK119Ub). TRIM37 also possesses anti-HIV activity. Mutation in TRIm37 causes an autosomal recessive prenatal-onset growth disorder, mulibrey nanism with dysmorphic features, cardiomyopathy, and hepatomegaly.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Fernando R Balestra et al.
Developmental cell, 25(6), 555-571 (2013-06-19)
Centrioles are essential for forming cilia, flagella, and centrosomes and are thus critical for a range of fundamental cellular processes. Despite their importance, the mechanisms governing centriole biogenesis remain incompletely understood. We performed a high-content genome-wide small-interfering-RNA-based screen to identify...
Azah A Tabah et al.
The Journal of general virology, 95(Pt 4), 960-967 (2013-12-10)
Trim 5α was the first member of the tripartite motif (TRIM) family of proteins that was identified to potently restrict human immunodeficiency virus type 1 (HIV-1) replication. The breadth of antiretroviral activity of TRIM family members is an active area...
Jukka Kallijärvi et al.
Experimental cell research, 308(1), 146-155 (2005-05-12)
Mulibrey nanism is an autosomal recessive prenatal-onset growth disorder characterized by dysmorphic features, cardiomyopathy, and hepatomegaly. Mutations in TRIM37 encoding a tripartite motif (TRIM, RING-B-box-coiled-coil)-family protein underlie mulibrey nanism. We investigated the ubiquitin ligase activity predicted for the RING domain...
Yun Li et al.
The Journal of pathology, 246(3), 366-378 (2018-07-26)
Non-small-cell lung cancer (NSCLC), in which the NF-κB pathway is constitutively activated, is one of the most common malignancies. Herein, we identify an E3 ubiquitin ligase, tripartite motif-containing 37 (TRIM37), participating in the K63 polyubiquitination of TRAF2, which is a...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.