SAB2900193

Sigma-Aldrich

Anti-BACE1 (AB1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):
bace, b-Secretase, asp2, aspartylprotease 2, APP beta-secretase
Pricing and availability is not currently available.

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

application(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BACE1(23621)

Immunogen

Synthetic 18 amino acid peptide from near N-terminus of human BACE1. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Rat (100%); Mouse, Bovine, Hamster, Panda (94%); Bat (89%); Elephant, Horse, Rabbit (83%).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Hazard Codes

Xn

Risk Statement

22

RIDADR

NONH for all modes of transport

Certificate of Analysis
Certificate of Origin
Chang Yang et al.
Neuropeptides, 48(4), 233-238 (2014-05-13)
Nerve growth factor (NGF) is an important nerve cell growth regulatory factor and has an indispensable role in the development, survival and regeneration of the cholinergic basal forebrain (CBF) neurons, and it has multiple targets when used for Alzheimer's Disease...
Jian Zhang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(45), 14919-14933 (2014-11-08)
Abnormal accumulation of β-amyloid (Aβ) is the major neuropathological hallmark of Alzheimer's disease (AD). However, the mechanisms underlying aberrant Aβ formation in AD remain unclear. We showed previously that inhibition of monoacylglycerol lipase (MAGL), the primary enzyme that metabolizes the...
Xiangmei Wu et al.
Molecular neurobiology, 50(3), 839-851 (2014-04-15)
Chronic cerebral hypoperfusion is associated with cognitive decline in aging and age-related neurodegenerative disease. Epigenetic mechanisms are involved in the maintenance of long-term hypoxia-adapted cellular phenotypes. In the present study, the epigenetic signatures such as DNA methylation and histone acetylation...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.