SAB3501047

Sigma-Aldrich

Anti-IL-36A antibody produced in rabbit

affinity isolated antibody

NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 17 kDa

species reactivity

human

concentration

1 mg/mL

application(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL36A(27179)

Immunogen

Antibody was raised against an 11 amino acid peptide near the carboxy terminus of human IL-36A.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501047.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Interleukin-36 (IL-36) ligands require processing for full agonist agonist (IL-36a, IL-36b, and IL-36g) or antagonist (IL-36Ra) activity
Towne JE, et al
The Journal of Biological Chemistry, 286, 42954-42602 (2011)
Solenne Vigne et al.
Blood, 120(17), 3478-3487 (2012-09-13)
The interleukin-1 (IL-1) superfamily of cytokines comprises a set of pivotal mediators of inflammation. Among them, the action of IL-36 cytokines in immune responses has remained elusive. In a recent study, we demonstrated a direct effect of IL-36 on immune...
D E Smith et al.
The Journal of biological chemistry, 275(2), 1169-1175 (2000-01-08)
We report here the cloning and characterization of four new members of the interleukin-1 (IL-1) family (FIL1delta, FIL1epsilon, FIL1zeta, and FIL1eta, with FIL1 standing for "Family of IL-1"). The novel genes demonstrate significant sequence similarity to IL-1alpha, IL-1beta, IL-1ra, and...
Jennifer E Towne et al.
The Journal of biological chemistry, 279(14), 13677-13688 (2004-01-22)
Interleukin 1 (IL-1) plays a prominent role in immune and inflammatory reactions. Our understanding of the IL-1 family has recently expanded to include six novel members named IL-1F5 to IL-1F10. Recently, it was reported that IL-1F9 activated NF-kappaB through the...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service