SAB3501050

Sigma-Aldrich

Anti-IL-36RN antibody produced in rabbit

affinity isolated antibody

NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 24 kDa

species reactivity

human, mouse

concentration

1 mg/mL

application(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL36RN(26525)

Immunogen

Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human IL-36RN.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501050.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Hal Blumberg et al.
The Journal of experimental medicine, 204(11), 2603-2614 (2007-10-03)
The interleukin (IL)-1 family members IL-1alpha, -1beta, and -18 are potent inflammatory cytokines whose activities are dependent on heterodimeric receptors of the IL-1R superfamily, and which are regulated by soluble antagonists. Recently, several new IL-1 family members have been identified....
R Debets et al.
Journal of immunology (Baltimore, Md. : 1950), 167(3), 1440-1446 (2001-07-24)
IL-1 is of utmost importance in the host response to immunological challenges. We identified and functionally characterized two novel IL-1 ligands termed IL-1delta and IL-1epsilon. Northern blot analyses show that these IL-1s are highly abundant in embryonic tissue and tissues...
D E Smith et al.
The Journal of biological chemistry, 275(2), 1169-1175 (2000-01-08)
We report here the cloning and characterization of four new members of the interleukin-1 (IL-1) family (FIL1delta, FIL1epsilon, FIL1zeta, and FIL1eta, with FIL1 standing for "Family of IL-1"). The novel genes demonstrate significant sequence similarity to IL-1alpha, IL-1beta, IL-1ra, and...
Jennifer E Towne et al.
The Journal of biological chemistry, 286(49), 42594-42602 (2011-10-04)
IL-36α, IL-36β, and IL-36γ (formerly IL-1F6, IL-1F8, and IL-1F9) are IL-1 family members that signal through the IL-1 receptor family members IL-1Rrp2 (IL-1RL2) and IL-1RAcP. IL-36Ra (formerly IL-1F5) has been reported to antagonize IL-36γ. However, our previous attempts to demonstrate...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.