SAB4100489

Sigma-Aldrich

Monoclonal Anti-CD4 antibody produced in mouse

clone CD4 -A409, culture supernatant

Synonym(s):
Anti-CD4mut
Pricing and availability is not currently available.

Quality Level

biological source

mouse

antibody form

culture supernatant

antibody product type

primary antibodies

clone

CD4 -A409, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 39.3 kDa

species reactivity

human

application(s)

western blot: suitable

isotype

IgG2a

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CD4(920)

Immunogen

CD4 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Kouichi Furugaki et al.
PloS one, 9(7), e101854-e101854 (2014-07-12)
Polyplex micelles have demonstrated biocompatibility and achieve efficient gene transfection in vivo. Here, we investigated a polyplex micelle encapsulating genes encoding the tumor-associated antigen squamous cell carcinoma antigen recognized by T cells-3 (SART3), adjuvant CD40L, and granulocyte macrophage colony-stimulating factor...
Min-Chul Kim et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(7), 1364-1374 (2014-03-05)
Current influenza vaccines do not provide good protection against antigenically different influenza A viruses. As an approach to overcome strain specificity of protection, this study demonstrates significantly improved long-term cross protection by supplementing split vaccines with a conserved molecular target...
Koichi Iwanaga et al.
Cancer research, 74(11), 3011-3019 (2014-06-01)
Compared with prostaglandin E2, which has an established role in cancer, the role of the COX metabolite prostaglandin D2 (PGD2) in chronic inflammation leading to tumorigenesis is uncertain. In this study, we investigated the role of PGD2 in colitis and...
Chiuhui Mary Wang et al.
The EMBO journal, 33(12), 1354-1364 (2014-05-21)
Organization of immune responses requires exchange of information between cells. This is achieved through either direct cell-cell contacts and establishment of temporary synapses or the release of soluble factors, such as cytokines and chemokines. Here we show a novel form...
Julie Lajoie et al.
The Journal of infectious diseases, 210(2), 319-329 (2014-01-15)
Unprotected intercourse and seminal discharge are powerful activators of the mucosal immune system and are important risk factors for transmission of human immunodeficiency virus (HIV). This study was designed to determine if female sex work is associated with changes in...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.