SAB4100595

Sigma-Aldrich

Monoclonal Anti-FCER2 antibody produced in mouse

clone FCER2-A577, culture supernatant

Synonym(s):
Anti-CLEC4J, Anti-FCE2, Anti-CD23, Anti-CD23A, Anti-IGEBF
NACRES:
NA.41

Quality Level

biological source

mouse

antibody form

culture supernatant

antibody product type

primary antibodies

clone

FCER2-A577, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 23.1 kDa

species reactivity

human

application(s)

western blot: suitable

isotype

IgG2a

conjugate

unconjugated

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FCER2(2208)

Immunogen

FCER2 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Saori Fukao et al.
Journal of immunology (Baltimore, Md. : 1950), 193(2), 635-644 (2014-06-18)
The rapid Ab responses observed after primary and secondary immunizations are mainly derived from marginal zone (MZ) and memory B cells, respectively, but it is largely unknown how these responses are negatively regulated. Several inhibitory receptors have been identified and...
Ryan Flynn et al.
Blood, 123(25), 3988-3998 (2014-05-14)
Chronic graft-versus-host disease (cGVHD) is a leading cause of morbidity and mortality after allogeneic hematopoietic stem cell transplantation. Having shown that germinal center (GC) formation and immunoglobulin deposition are required for multiorgan system cGVHD and associated bronchiolitis obliterans syndrome (BOS)...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.