SAB4301816

Sigma-Aldrich

Anti-UBE2W

affinity isolated antibody

Synonym(s):
UBC-16

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, rat

application(s)

immunohistochemistry: 1: 100-250
western blot: 1:500-1:2000

accession no.

BC016326

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... UBE2W(55284)

Immunogen

Full length fusion protein

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.4 PBS, 0.05% NaN3, 40% Glycerol.

storage_class_code

12 - Non Combustible Liquids

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service