SAB4500613
Anti-Cytochrome P450 27A1 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
25-hydroxylase, 5-β-cholestane-3-α,7-α,12-α-triol 27-hydroxylase, CP27A, Cytochrome P-450C27/25, Cytochrome P450 27, mitochondrial
Select a Size
$392.00
Select a Size
About This Item
Skip To
$392.00
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 60 kDa
species reactivity
human
concentration
~1 mg/mL
technique(s)
ELISA: 1:10000
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CYP27A1(1593)
1 of 4
This Item | SAB4500609 | SAB4500585 | SAB4500586 |
---|---|---|---|
conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
General description
Immunogen
Immunogen Range: 101-150
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service