SAB4502429

Sigma-Aldrich

Anti-BCA3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):
Breast cancer-associated gene 3 protein, PKA-interacting protein, AKIP1
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 23 kDa

species reactivity

human

concentration

~1 mg/mL

application(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

conjugate

unconjugated

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BCA3(56672)

General description

Anti-BCA3 Antibody detects endogenous levels of total BCA3 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human BCA3.

Immunogen Range: 111-160

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

WGK Germany

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Yanmei Cui et al.
Oncogene, 38(27), 5516-5529 (2019-04-03)
The early recurrence of hepatocellular carcinoma (HCC) is the main obstacle for long-term survival of patients. Wnt/β-catenin signaling has been involved in the development and progression of HCC. However, the molecular changes that link Wnt/β-catenin activation and HCC early recurrence...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service