SAB4502879

Sigma-Aldrich

Anti-STAT5A antibody produced in rabbit

affinity isolated antibody

Synonym(s):
STA5A, STAT5, Mammary gland factor, MPF, MGF
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 89 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

application(s)

ELISA: 1:1000
western blot: 1:500-1:1000

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... STAT5A(6776)

General description

Anti-STAT5A Antibody detects endogenous levels of total STAT5A protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human STAT5A/B.

Immunogen Range: 251-300

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

storage_class_code

12 - Non Combustible Liquids

WGK Germany

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Alex Man Lai Wu et al.
American journal of physiology. Endocrinology and metabolism, 307(7), E596-E610 (2014-08-15)
The multidrug resistance efflux transporter ATP-binding cassette subfamily G member 2 (ABCG2) is not only overexpressed in certain drug-resistant cancers but is also highly expressed in the mammary gland during lactation, carrying xenobiotics and nutrients into milk. We sought to...
Liting Xue et al.
Blood, 124(3), 426-436 (2014-06-05)
Acute myeloid leukemia (AML) results from the activity of driver mutations that deregulate proliferation and survival of hematopoietic stem cells (HSCs). The fusion protein CBFβ-SMMHC impairs differentiation in hematopoietic stem and progenitor cells and induces AML in cooperation with other...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service