SAB4503404

Sigma-Aldrich

Anti-IKK-γ antibody produced in rabbit

affinity isolated antibody

Synonym(s):
NEMO, IkB kinase subunit γ, I-κ-B kinase γ, Inhibitor of nuclear factor κ-B kinase subunit γ, NF-κ-B essential modifier
MDL number:
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human

concentration

~1 mg/mL

application(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IKBKG(8517)

General description

Anti-IKK-γ Antibody detects endogenous levels of total IKK-γ protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human IKK-gamma.

Immunogen Range: 51-100

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

storage_class_code

12 - Non Combustible Liquids

WGK Germany

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Xiao-Hui Zou et al.
Virology, 468-470, 388-396 (2014-09-23)
The human adenovirus (HAdV) early protein E1B55K interacts with E4orf6 to form an E3 ubiquitin ligase complex, which plays key roles in virus replication. To illustrate the reason for the fastidiousness of HAdV-41 in 293 cells, interaction between heterotypic E1B55K...
Jingnan Sun et al.
Nucleic acids research, 42(15), 9588-9601 (2014-08-06)
Dysregulation of the insulin-like growth factor type I receptor (IGF1R) has been implicated in the progression and therapeutic resistance of malignancies. In acute myeloid leukemia (AML) cells, IGF1R is one of the most abundantly phosphorylated receptor tyrosine kinases, promoting cell...
Manuraj Pandey et al.
Toxicology and applied pharmacology, 280(2), 296-304 (2014-08-30)
In the present study, we showed the correlation of EZH2, SUV39H1 or G9a expression and histone modifications with the urethane induced mouse lung tumorigenesis in the presence or absence of antitumor agent, inositol hexaphosphate (IP6). Tumorigenesis and the molecular events...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service