MilliporeSigma
All Photos(2)

SAB4504624

Sigma-Aldrich

Anti-phospho-MAP2K7 (pThr275) antibody produced in rabbit

affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 47 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAP2K7(5609)

Immunogen

The antiserum was produced against synthesized peptide derived from human MAP2K7 around the phosphorylation site of Thr275.

Immunogen Range: 241-290

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Masahiro Kawahara et al.
Biotechnology journal, 9(7), 954-961 (2014-01-01)
Since receptor tyrosine kinases (RTKs) control various cell fates in many types of cells, mimicry of RTK functions is promising for artificial control of cell fates. We have previously developed single-chain Fv (scFv)/receptor chimeras named signalobodies that can mimic receptor
Armelle-Natsuo Takeda et al.
The EMBO journal, 33(16), 1784-1801 (2014-07-01)
Mitogen-activated protein kinases (MAPKs) are highly conserved protein kinase modules, and they control fundamental cellular processes. While the activation of MAPKs has been well studied, little is known on the mechanisms driving their inactivation. Here we uncover a role for
Akriti Kharbanda et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(21), 5423-5434 (2014-09-06)
Non-small cell lung cancers (NSCLC) that express EGF receptor with activating mutations frequently develop resistance to EGFR kinase inhibitors. The mucin 1 (MUC1) heterodimeric protein is aberrantly overexpressed in NSCLC cells and confers a poor prognosis; however, the functional involvement
K Sanjana P Devi et al.
Biochimica et biophysica acta, 1840(9), 2794-2805 (2014-05-09)
Newer strategies for augmenting immune responses of pharmacologically active glucans may serve to improve the medicinal potential of these biomolecules. With this aim, the present work was focused on generating targeted high molecular size glucan particles with magnified immune response
Baosen Jia et al.
Journal of virology, 89(4), 2041-2051 (2014-11-28)
Hepatitis B, which caused by hepatitis B virus (HBV) infection, remains a major health threat worldwide. Hepatic injury and regeneration from chronic inflammation are the main driving factors of liver fibrosis and cirrhosis in chronic hepatitis B. Proinflammatory tumor necrosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service