SAB4700678

Sigma-Aldrich

Monoclonal Anti-CD23-PE antibody produced in mouse

clone EBVCS-5, purified immunoglobulin, buffered aqueous solution

Synonym(s):
Anti-FcepsilonRII
Pricing and availability is not currently available.

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

EBVCS-5, monoclonal

form

buffered aqueous solution

species reactivity

human

application(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

conjugate

phycoerythrin (R-PE) conjugate

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... FCER2(2208)

General description

The mouse monoclonal antibody EBVCS-5 recognizes an epitope located in the stalk region of human low affinity IgE receptor (CD23) between the 37 and 25 kDa cleavage sites.

Immunogen

EBV-transformed human cells

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.