SAB4800002

Sigma-Aldrich

Imprint® Monoclonal Anti-5-hydroxymethylcytosine antibody produced in rat

affinity isolated antibody, buffered aqueous solution

Synonym(s):
Anti-5-hmC, Anti-5-hydroxymethylC

biological source

rat

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

buffered aqueous solution

species reactivity

mouse, human

concentration

~1.6 mg/mL

application(s)

indirect ELISA: suitable
methylated DNA immunoprecipitation (MeDIP): suitable

conjugate

unconjugated

shipped in

dry ice

storage temp.

−20°C

Physical form

Supplied at 1.6 mg/mL in PBS, 0.05% sodium azide.

Legal Information

Imprint is a registered trademark of Sigma-Aldrich Co. LLC

RIDADR

NONH for all modes of transport

WGK Germany

nwg

Certificate of Analysis
Certificate of Origin
Articles
Epigenetic modifications are thought to occur through two key interconnected processes—DNA methylation and the covalent modification of histones.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.