SAB4800013

Sigma-Aldrich

Imprint® Anti-TIP5 antibody produced in rabbit

whole antiserum, buffered aqueous solution

Synonym(s):
Anti-BAZ2A, Anti-WALp3, Anti-TIP5

biological source

rabbit

Quality Level

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~211 kDa

species reactivity

mouse, human

application(s)

ChIP: suitable
western blot: suitable

conjugate

unconjugated

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BAZ2A(11176)

Physical form

Supplied as whole antiserum containing 0.05% sodium azide.

Legal Information

Imprint is a registered trademark of Sigma-Aldrich Co. LLC

RIDADR

NONH for all modes of transport

WGK Germany

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Articles
Epigenetic modifications are thought to occur through two key interconnected processes—DNA methylation and the covalent modification of histones.
Read More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.