SAB4800046

Sigma-Aldrich

Imprint® Anti-H4K8ac antibody produced in rabbit

purified antibody, buffered aqueous solution

Synonym(s):
Anti-Histone H4 (acetyl K8)

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~12 kDa

species reactivity

human

concentration

~1.23 mg/mL

application(s)

ChIP: suitable
indirect ELISA: suitable
western blot: suitable

conjugate

unconjugated

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... H4F3(3023)

Physical form

Supplied at 1.23 mg/mL in PBS, 0.05% sodium azide., 0.05% ProClin 300.

Legal Information

Imprint is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

RIDADR

NONH for all modes of transport

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.